Product Image
Thermo Scientific™
CPVC Protected Salt Bridge
CPVC protected salt bridge 1pc for differential pH electrodes AnalogPlus FP SP and DataStick
Have Questions?
Número do catálogo SBS03-1
Preço (BRL)
-
CPVC protected salt bridge 1pc for differential pH electrodes AnalogPlus FP SP and DataStick
AquaSensors AnalogPlus™ pH 1.0 Sensor
  • 1 inch NPT mounting
  • Differential pH sensor designed for continuous use in the most demanding industrial applications
  • Replaceable salt bridge extends sensor life 
  • Offered with NTC300 or PT1000 temperature elements
  • Electrode protection options
  • Available in choice of materials for a wide range of applications
Market/Applications
  • Wastewater treatment
  • Pharmaceutical
  • Chemical & petrochemical
  • High purity water
  • Food processing
  • Metal finishing
Especificações
TypepH Electrode Spare
Unit SizeEach