SureClick™ Antibody Labeling Kit
Invitrogen™

SureClick™ Antibody Labeling Kit

Invitrogen SureClick Antibody Labeling Kits enable fast, precise, and reproducible conjugation of human or rabbit IgG antibodies using trusted click chemistry. Designed for spatial biology researchers and biotherapeutic developers, SureClick kits attach an average of 2 payloads per antibody at the Fc region.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
SC0023 x 1 mg
SC0013 x 100 μg
SC0035 mg
Número de catálogo SC002
Precio (MXN)
-
Cantidad:
3 x 1 mg
Pedido a granel o personalizado

Covalent, Fc-targeted antibody conjugation in just two hours

Invitrogen SureClick Antibody Labeling Kits enable fast, precise, and reproducible conjugation of human or rabbit IgG antibodies using trusted click chemistry. Designed for spatial biology researchers and biotherapeutic developers, SureClick kits attach an average of 2 payloads per antibody at the Fc region, preserving antigen binding and Fc receptor (FcR) affinity.

SureClick is an exceptional solution for attaching oligonucleotides to antibodies, making it ideal for spatial biology workflows, immuno-PCR, and next-generation antibody-based detection platforms that require precise and reproducible antibody–oligo conjugation.

Achieve consistent, covalent antibody labeling with a simplified workflow amenable to automation.

What is SureClick antibody labeling?

SureClick is a site-specific antibody conjugation technology that covalently attaches payloads—including fluorophores, functional tags, and oligonucleotides—to the Fc region of IgG antibodies using click chemistry. Unlike conventional random labeling approaches, SureClick:

  • Targets the Fc domain
  • Preserves Fab antigen-binding regions
  • Maintains Fc glycosylation integrity
  • Enables reproducible degree of labeling (DOL)
  • Supports consistent antibody–oligonucleotide conjugation

Key features and benefits:

Precise, reproducible labeling

  • Site-specific Fc conjugation
  • Consistent attachment of ~2 payloads per antibody
  • Reduced heterogeneity compared to conventional methods

Rapid two-hour workflow

  • ~10 minutes hands-on time
  • 90-minute protocol
  • Simple protocol using a single Fc-targeting component

Covalent and stable conjugation

  • Trusted click chemistry
  • Robust linkage for downstream assays, spatial biology, and antibody–oligo workflows

Preserves antibody function

  • Confidence that antigen binding is unchanged
  • Maintains Fc receptor interactions

Automation-ready

  • Spin-column purification
  • Minimal components
  • Suitable for high-throughput workflows

How SureClick antibody labeling works

  1. Add SureClick Fc-targeting reagent to your IgG antibody
  2. Purify using included Zeba™ spin columns
  3. Add click chemistry payload of choice (e.g., fluorophores, oligonucleotides, functional tags)

The result is a site-specific, covalently labeled antibody with consistent performance, including highly controlled antibody–oligonucleotide conjugates.

Applications

  • Spatial biology
  • Antibody–oligonucleotide conjugation
  • Multiplex biomarker detection
  • Antibody-drug conjugate (ADC) development
  • Biotherapeutic characterization
  • Flow cytometry
  • Internalization studies
  • Biomarker validation
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Para utilizar con (aplicación)Antibody Production and Labeling
Para utilizar con (equipo)Benchtop Centrifuge
FormatoLabeling Kit
Tiempo de incubación70 to 90 min.
Método de etiquetadoClick Chemistry
Escala de etiquetadoApprox. 2°
Línea de productosSureClick™
Tipo de productoAntibody Labeling Kit
Cantidad3 x 1 mg
Fracción reactivaAzide
Condiciones de envíoWet Ice
EspecieHuman or Rabbit
Suficiente paraLabeling 3 Antibodies
Reactividad químicaAzide
Labeling TargetAntibodies
Etiqueta o tinteAzide
SolubilidadAqueous Buffer
SeparadorMid-length
Unit SizeEach
Contenido y almacenamiento
SureClick™ Fc Targeting Reagent
2°C to 8°C

Preguntas frecuentes

What antibody types are compatible?

SureClick labeling is designed for human and rabbit IgG antibodies.

How long does SureClick labeling take?

The full protocol takes approximately 90 minutes with about 10 minutes of hands-on time.

Does SureClick labeling affect antigen binding?

No. The SureClick reaction targets the Fc region, preserving the Fab antigen-binding domain and maintaining Fc receptor interactions..

Is SureClick labeling suitable for attaching oligonucleotides to antibodies?

Yes. SureClick labeling is an exceptional method for antibody–oligonucleotide conjugation. Its Fc-specific, site-controlled labeling enables reproducible and precise attachment of oligonucleotides while preserving antigen binding and antibody functionality.

How many labels does the SureClick reagent attach per antibody?

The SureClick reagent consistently attaches approximately two payloads per antibody.