Product Image
Thermo Scientific™
AnalogPlus Contacting Conductivity Sensor
AnalogPlus contacting conductivity sensor CPVC body titanium electrodes K=0.1 nominal cell constant 500uS/cm nominal range 9-metre cable
Have Questions?
Catalog NumberConnector Types
SC21A130Not Stated
Catalog number SC21A130
Price (MXN)
-
Connector Types:
Not Stated
AnalogPlus contacting conductivity sensor CPVC body titanium electrodes K=0.1 nominal cell constant 500uS/cm nominal range 9-metre cable
AnalogPlus contacting conductivity sensor CPVC body titanium electrodes K=0.1 nominal cell constant 500uS/cm nominal range 9-metre cable.
Specifications
Sensor TypesConductivity
Cell Constant0.1
Connector TypesNot Stated
MaterialCPVC Body, Titanium Electrodes
Measurement Ranges500 μS/cm nominal range
TypeConductivity Sensor
Unit SizeEach