Jurkat Genomic DNA
Thermo Scientific™

Jurkat Genomic DNA

Thermo Scientific Jurkat Genomic DNA is human male genomic DNA from Jurkat cells. Genomic DNA was isolated from Jurkat (humanRead more
Have Questions?
Catalog NumberQuantity
SD111115 μg
Catalog number SD1111
Price (USD)
104.65
Online Exclusive
112.00
Save 7.35 (7%)
15 µg
Quantity:
15 μg
Price (USD)
104.65
Online Exclusive
112.00
Save 7.35 (7%)
15 µg
Thermo Scientific Jurkat Genomic DNA is human male genomic DNA from Jurkat cells. Genomic DNA was isolated from Jurkat (human acute T-cell leukemia) cells and grown to confluency in RPMI media supplemented with 10% bovine fetal serum. Isolated genomic DNA is dissolved in TE buffer.

Higlights
• Pure—DNA free from inhibitors, proteins, and RNA
• Convenient—ready to use 100 ng/μL concentration
• Compatible with downstream applications

Applications

• Genomic DNA library construction
• PCR and Southern blotting

Also used as controls in the following:
• Bisulfite sequencing
• Combined Bisulfite Restriction Analysis (COBRA)
• Methylation-sensitive Single-Nucleotide Primer Extension (Ms-SNuPE)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferTE Buffer
Concentration100 ng/μL
For Use With (Application)Sequencing
FormLiquid
Includes[other]
Product TypeJurkat Genomic DNA
Quantity15 μg
SpeciesHuman
FormatTube
Unit Size15 µg