Thermo Scientific™ InSight sensors work with the mote transmitters, relaying data to the InSight Cloud service.
Have Questions?
Change view
Catalog Number
Description
SENS-DOOR-1001
Door access sensor
SENS-RTD-1002
RTD: RE style RTD assembly; single wire-wound 100 ohm platinum element, Class B tolerance; 14 foot PFA
2 Options
Catalog number SENS-DOOR-1001
Price (KRW)
-
Description:
Door access sensor
Thermo Scientific™ InSight sensors relay data from the motes on your wireless account to the InSight Cloud service. In the event of internet failure, Insight Cloud will notify IT and the InSight link will log sensor data for data backfill on recovery.
Door access sensor
RTD sensor
Warranty varies by country, please contact us for details
Not available outside North America
Specifications
DescriptionDoor access sensor
For Use With (Application)Remote Monitoring, Asset Management Solution