Owl™ SGC Sequencing Gel Casters
Actual product may vary
Owl™ SGC Sequencing Gel Casters
Owl™ SGC Sequencing Gel Casters
Thermo Scientific™

Owl™ SGC Sequencing Gel Casters

Cast bubble-free gels in less than three minutes with Thermo Scientific™ Owl™ SGC Sequencing Gel Casters.
Have Questions?
카탈로그 번호길이(미터법)
SGC-2102 cm
카탈로그 번호 SGC-2
제품 가격(KRW)
-
길이(미터법):
102 cm
Cast bubble-free gels in less than three minutes with Thermo Scientific™ Owl™ SGC Sequencing Gel Casters.
Features:
  • No taping or special assembly required
  • Exposure to hazardous acrylamide is decreased, reducing cleanup time
  • Available in two lengths: 48 or 65.5cm
  • Fully adjustable to accommodate glass plates from 20 to 42cm wide

Includes:

  • Adjustable casting platform
  • Width adjustment knobs
  • 12 binder clamps
사양
치수(길이 x 폭 x 높이)40 x 8.25 x 4.75 in. (102 x 21 x 12.2 cm)
높이(영국식 단위)4.75 in.
높이(미터법)12.2 cm
길이(영국식 단위)40 in.
길이(미터법)102 cm
길이(미터법) 젤20 to 42 cm
폭(영국식 단위)8.25 in.
폭(미터법)21 cm
폭(미터법) 젤48 cm
용도 (장비)Owl SGC Sequencing System
젤 크기10 x 10 cm
제품라인Owl
유형Gel Caster
Unit SizeEach