2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 2 ports, MPC insert connectors
2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 2 ports, MPC insert connectors
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 2 ports, MPC insert connectors

The small- and medium-sized Thermo Scientific™ Labtainer™ BioProcess Containers (BPCs) range in size from 100 mL to 2 L. TheseLeia mais
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoCapacity (Metric)
SH00019I
conhecido também como SH00019-I
500 mL
SH00017I
conhecido também como SH00017-I
100 mL
SH00018I
conhecido também como SH00018-I
250 mL
SH00020I
conhecido também como SH00020-I
1 L
SH00021I
conhecido também como SH00021-I
2 L
Número do catálogo SH00019I
conhecido também como SH00019-I
Preço (BRL)
-
Capacity (Metric):
500 mL
The small- and medium-sized Thermo Scientific™ Labtainer™ BioProcess Containers (BPCs) range in size from 100 mL to 2 L. These bags feature ¼-inch ID tubing, making them ideal for smaller flow applications. The MPC insert connectors are easy to use and offer secure fluid handling. The 1 L Thermo Scientific™ BPC 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL ethyl vinyl acetate film, and includes 2 ports (2 MPC insert + barb and cap).

Features of the system include:
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications include:
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquid
• Collecting chromatography feed and fraction
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors
• Filters (used for bioburden reduction)—sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers and platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Especificações
Dimensions18.4 cm x 12.4 cm (7.25 in. X 4.88 in.)
Outer PackagingTwo 4-mil. poly bags
Product LineLabtainer
Quantity500 mL, 50 per case
RegulatorycGMP
Shipping ConditionRoom Temperature
Sterilization MethodGamma Irradiated
Capacity (Metric)500 mL
Film DescriptionASI 28
MaterialEVA
Port DetailMPC Insert Connector
Tubing StyleEVA
Unit Size500 mL
Conteúdo e armazenamento
Store at room temperature.