2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 1 port, Luer Lock connector
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 1 port, Luer Lock connector

The 5 L Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, aMás información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)
SH00041I
también denominado SH00041-I
10 L
SH00040I
también denominado SH00040-I
5 L
SH00042I
también denominado SH00042-I
20 L
Número de catálogo SH00041I
también denominado SH00041-I
Precio (MXN)
-
Capacidad (métrico):
10 L
The 5 L Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 1 port (1 luer body + cap).

Features of the system include:
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications include:
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquid
• Collecting chromatography feed and fraction
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors
• Filters (used for bioburden reduction)—sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers and platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Especificaciones
Dimensiones56.5 cm x 32.4 cm (22.25 in. X 12.75 in.)
Embalaje exteriorTwo 4-mil. poly bags
Línea de productosLabtainer
Cantidad10 L, 25 per case
ReglamentacióncGMP
Condiciones de envíoTemperatura ambiente
Método de esterilizaciónGamma Irradiated
Capacidad (métrico)10 L
Film DescriptionASI 28
MaterialEVA
Port DetailMPC Insert Connector
Tubing StyleEVA
Unit Size10 L
Contenido y almacenamiento
Almacenar a temperatura ambiente.