HyPerforma™ 50 to 500L Tank Liners, 100L
Thermo Scientific™
HyPerforma™ 50 to 500L Tank Liners, 100L
The 100L Thermo Scientific™ tank liner system with top drain is composed of CX3-9 single web, multi-layer plastic film, withRead more
Have Questions?
Catalog NumberQuantity
SH3064702
Catalog number SH3064702
Price (USD)
-
The 100L Thermo Scientific™ tank liner system with top drain is composed of CX3-9 single web, multi-layer plastic film, with dimensions optimized for Thermo Scientific™ drums and commonly used industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
Features:
• Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
• Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications of the Thermo Scientific tank liner:
• Removes the need for tank cleaning and helps reduce cycle times.
• Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
Specifications
For Use With (Application)Handling Fluids in Bioproduction Application
For Use With (Equipment)SV50076.03
Outer PackagingPackages are placed in strong cardboard cartons with product and lot identification.
Product LineHyPerforma
Product TypeTank Liner
Protective Inner PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area.
SterilitySterile
Sterilization MethodIrradiated
Capacity (Metric)100 L
Film DescriptionCX3-9
MaterialPlastic