HyPerforma™ 50 to 500L Tank Liners, 50L
Thermo Scientific™
HyPerforma™ 50 to 500L Tank Liners, 50L
The 50L Thermo Scientific™ tank liner system with top drain is composed of CX3-9 single web, multi-layer plastic film, with詳細を見る
製品番号(カタログ番号)Quantity
SH3064704
または、製品番号SH30647.04
製品番号(カタログ番号) SH3064704
または、製品番号SH30647.04
価格(JPY)
-
The 50L Thermo Scientific™ tank liner system with top drain is composed of CX3-9 single web, multi-layer plastic film, with dimensions optimized for Thermo Scientific™ drums and commonly used industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
Features:
• Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
• Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications of the Thermo Scientific tank liner:
• Removes the need for tank cleaning and helps reduce cycle times.
• Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
仕様
使用対象 (装置)SV30111
外部包装Packages are placed in strong cardboard cartons with product and lot identification.
製品ラインHyPerforma
製品タイプTank Liner
保護用内部包装Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area.
無菌性Sterile
滅菌方法Irradiated
容量(メートル法)50 L
Film DescriptionCX3-9
材料Plastic