Productainer™ BioProcess Container (BPC), 100 L with 4 TopPorts and 1 Bottom Dispense, 3D Single-Use BPC
Thermo Scientific™

Productainer™ BioProcess Container (BPC), 100 L with 4 TopPorts and 1 Bottom Dispense, 3D Single-Use BPC

The Thermo Scientific™ Productainer™ BioProcess Container (BPC), 100 L 3D system is composed of CX5-14 single web, multi-layer plastic filmRead more
Have Questions?
Catalog NumberQuantity
SH3065202
also known as SH30652.02
100 L
Catalog number SH3065202
also known as SH30652.02
Price (MXN)
-
Quantity:
100 L
The Thermo Scientific™ Productainer™ BioProcess Container (BPC), 100 L 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 4 top ports and 1 bottom port. The 3D square tube design with bottom port dispense is for use with conical, bottom-drain drums.

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All 3D BPCs are constructed in an ISO 7 cleanroom under cGMP conditions.
• All 3D BPCs are designed to fit the full range of support containers, both square and cylindrical – from 50 to 2000 L.

Applications of the Thermo Scientific 3D BPC include:
• Hydration and sterilization of process buffers, liquids, and culture media.
• Chromatography feed and fraction collection.
• Storage and transport of bulk drug product and bulk drug precursors.
• Harvest from and feed into bioreactors and fermentors.
• Dispensing, packaging, and storage of cell culture media, buffers and process liquids.

Customize standard BPC systems for optimized single-use technology. Choose from the industry’s largest component library – with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
• Filters—used for bioburden reduction; sterilization grade filters and vent filters.
• Tubing—thermoplastic elastomers, platinum-cured silicone.
• Sample ports—septum-style and needle-free ports.
• Clamps—sliders, pinch, and crimp bands.

Specifications
Film DescriptionCX5-14
For Use With (Equipment)SV50517.05
Outer PackagingPackages placed in strong cardboard cartons with product and lot identification
Port DetailBottom Port: 1 x 0.38 in., Top Ports: 1 x 0.38 in. 2 x 0.25 in.
Product LineProductainer
Protective Inner PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity100 L
Sterilization MethodGamma Irradiated
Tubing Line 1Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 121.9 cm (48 in.) dip tube ID x OD: 9.5 x 15.9 mm (0.38 in. x 0.63 in.) with dip tube length: 30.5 cm (12 in.) Top Port
Tubing Line 2Line 2. Insert luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 in. x 0.38 in.) Top Port
Tubing Line 3Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 in. x 0.38 in.) Top Port
Tubing Line 4Line 4. Plugged, no tubing
Tubing Line 5Line 5. MPC body, polycarbonate Tubing: silicone, length: 121.9 cm (48 in.) ID x OD: 9.5 x 15.9 mm (0.38 in. x 0.63 in.) Bottom port
Capacity (Metric)100 L
Unit Size100 L