HyPerforma™ Smartainer™ 3D BPC, 1000L; 5 Ports
Thermo Scientific™

HyPerforma™ Smartainer™ 3D BPC, 1000L; 5 Ports

The 1000L Thermo Scientific™ large-volume BPC, 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 5Más información
Have Questions?
Número de catálogoCantidad
SH3065205
también denominado SH30652.05
1000 L
Número de catálogo SH3065205
también denominado SH30652.05
Precio (MXN)
-
Cantidad:
1000 L
The 1000L Thermo Scientific™ large-volume BPC, 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 5 top ports (MPC insert, male luer lock, female luer lock, plugged), 1 bottom drain (MPC body).

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All 3D BPCs are constructed in an ISO 7 cleanroom under cGMP conditions.
• All 3D BPCs are designed to fit the full range of support containers, both square and cylindrical – from 50 to 2000L.

Applications of the Thermo Scientific 3D BPC include:
• Hydration and sterilization of process buffers, liquids, and culture media.
• Chromatography feed and fraction collection.
• Storage and transport of bulk drug product and bulk drug precursors.
• Harvest from and feed into bioreactors and fermentors.
• Dispensing, packaging, and storage of cell culture media, buffers and process liquids.

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library – with over 20,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
Filters—used for bioburden reduction; sterilization grade filters and vent filters.
Tubing—thermoplastic elastomers, platinum-cured silicone.
Sample ports—septum-style and needle-free ports.
Clamps—sliders, pinch, and crimp bands.
Especificaciones
Descripción de la películaCX5-14
Para utilizar con (equipo)SV50160.03
Embalaje exteriorPaquetes colocados en cajas de cartón resistentes con identificación de productos y lotes
Detalle del puertoBottom Port: 1 x 0.38 in., Top Ports: 1 x 0.38 in. 2 x 0.25 in.
Línea de productosHyPerforma, Smartainer
Embalaje interior de protecciónPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Cantidad1000 L
Método de esterilizaciónGamma Irradiated
Línea de tuberías 1Line 1. MPC insert, polycarbonate, Tubing: TPE, length: 121.9 cm (48 in.) dip tube ID x OD: 9.5 x 15.9 mm (0.38 in. x 0.63 in.) with dip tube length: 30.5 cm (12 in.) Top Port
Línea de tuberías 2Line 2. Insert luer lock connection, polypropylene Tubing: TPE, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 in. x 0.38 in.) Top Port
Línea de tuberías 3Line 3. Body luer lock connection, polypropylene Tubing: TPE, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 in. x 0.38 in.) Top Port
Línea de tuberías 4Line 4. Plugged, no tubing
Línea de tuberías 5Line 5. MPC body, polycarbonate Tubing: silicone, length: 121.9 cm (48 in.) ID x OD: 9.5 x 15.9 mm (0.38 in. x 0.63 in.) Bottom port
Capacidad (métrico)1000 L
Tubing StyleTPE
Unit Size1000 L