Labtainer™ BioProcess Container (BPC), 500 mL with 2 Ports, Luer Lock, 2D BPC
Thermo Scientific™

Labtainer™ BioProcess Container (BPC), 500 mL with 2 Ports, Luer Lock, 2D BPC

The 500 mL Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single-web, multi-layer film, and includes 2 edgeRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Tubing Style
SH3065814
also known as SH30658.14
500 mL12 in. of 1/4 in. Tubing
SH30657.14500 mLNo Tubing
Catalog number SH3065814
also known as SH30658.14
Price (MXN)
-
Capacity (Metric):
500 mL
Tubing Style:
12 in. of 1/4 in. Tubing
The 500 mL Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single-web, multi-layer film, and includes 2 edge ports with luer lock body and insert connections.

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All Labtainers are constructed in an ISO 7 cleanroom under cGMP conditions.

Applications of the Thermo Scientific Labtainer BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors.
• Buffer, culture media, and process liquid preparation and storage.
• Chromatography feed and fraction collection.
• Harvest, storage and transport of bulk drug product and bulk drug precursors.

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library – with over 20,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
Filters—used for bioburden reduction; sterilization grade filters and vent filters.
Tubing—thermoplastic elastomers, platinum-cured silicone.
Sample ports—septum-style and needle-free ports.
Clamps—sliders, pinch, and crimp bands.

Specifications
Dimensions26.4 cm x 17.3 cm (10.4 in. x 6.8 in.)
Outer PackagingPackages placed in strong cardboard cartons with product and lot identification
Product LineLabtainer
Protective Inner PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity500 mL, 10 per case
Sterilization MethodGamma Irradiated
Tubing Line 1Line 1. Body luer lock connection,polypropylene Tubing: C-Flex, length: 30 cm (12 in.) Id x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Tubing Line 2Line 2. Insert luer lock connection, polypropylene Tubing: C-Flex, length: 30 cm (12 in.) ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Capacity (Metric)500 mL
Film DescriptionCX5-14
Port DetailEdge Ports: 2 x 0.25 in.
Tubing Style12 in. of 1/4 in. Tubing
Unit Size500 mL