Labtainer™ BioProcess Container (BPC), 1L with 2 Ports, Luer Lock and MPC Insert, 2D BPC
Thermo Scientific™

Labtainer™ BioProcess Container (BPC), 1L with 2 Ports, Luer Lock and MPC Insert, 2D BPC

The 1 L Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single-web, multi-layer film, and includes 2 edge詳細を見る
製品番号(カタログ番号)数量
SH3066215
または、製品番号SH30662.15
1 L, 10 per case
製品番号(カタログ番号) SH3066215
または、製品番号SH30662.15
価格(JPY)
-
数量:
1 L, 10 per case
The 1 L Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single-web, multi-layer film, and includes 2 edge ports with luer lock body and MPC insert.

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All Labtainers are constructed in an ISO 7 cleanroom under cGMP conditions.

Applications of the Thermo Scientific Labtainer BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors.
• Buffer, culture media, and process liquid preparation and storage.
• Chromatography feed and fraction collection.
• Harvest, storage and transport of bulk drug product and bulk drug precursors.

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library – with over 20,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
Filters—used for bioburden reduction; sterilization grade filters and vent filters.
Tubing—thermoplastic elastomers, platinum-cured silicone.
Sample ports—septum-style and needle-free ports.
Clamps—sliders, pinch, and crimp bands.

仕様
寸法29.7 cm x 20.1 cm (11.7 in. x 7.9 in.)
外部包装Packages placed in strong cardboard cartons with product and lot identification
製品ラインLabtainer
保護用内部包装Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
数量1 L, 10 per case
滅菌方法Gamma Irradiated
配管ライン1Line 1. Body luer lock connection,polypropylene Tubing: C-Flex, length: 30 cm (12 in.) Id x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
配管ライン2Line 2. MPC insert, polycarbonate, No Tubing
Film DescriptionCX5-14
Port DetailEdge Ports: 2 x 0.25 in.
Unit Size1 L