Labtainer™ BioProcess Container (BPC), 200 L with 3 Ports, 2D BPC
Product Image

Labtainer™ BioProcess Container (BPC), 200 L with 3 Ports, 2D BPC

Have Questions?
Catalog number SH3066703
Price (USD)
-
Request A Quote
The Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 200 L 2D pillow design is composed of CX5-14 single web, multi-layer plastic film and includes 3 panel ports.

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All Labtainers are constructed in an ISO 7 cleanroom under cGMP conditions.

Applications of the Thermo Scientific Labtainer BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors.
• Buffer, culture media, and process liquid preparation and storage.
• Chromatography feed and fraction collection.
• Harvest, storage and transport of bulk drug product and bulk drug precursors.

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library – with over 20,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
Filters—used for bioburden reduction; sterilization grade filters and vent filters.
Tubing—thermoplastic elastomers, platinum-cured silicone.
Sample ports—septum-style and needle-free ports.
Clamps—sliders, pinch, and crimp bands.

Specifications
Unit Size200 L