Mixtainer BioProcess Container (BPC), 50L, for HyPerforma™ Mixtainer™ Systems
Thermo Scientific™

Mixtainer BioProcess Container (BPC), 50L, for HyPerforma™ Mixtainer™ Systems

The 50L Thermo Scientific™ HyPerforma™ Mixtainer™ BioProcess Container (BPC) is composed of CX5-14 single web, multi-layer plastic film with 4Read more
Have Questions?
Catalog NumberQuantity
SH3068704
also known as SH30687.04
50 L
Catalog number SH3068704
also known as SH30687.04
Price (USD)
-
Quantity:
50 L
The 50L Thermo Scientific™ HyPerforma™ Mixtainer™ BioProcess Container (BPC) is composed of CX5-14 single web, multi-layer plastic film with 4 top ports. The complete Thermo Scientific™ Mixtainer™ System is an integrated, single-use sterile unit for mixing of cell culture media, and other process liquids such as buffers, reagents and bulk drug precursors or products. The system is available in 50, 100, and 200L unit volumes and is recommended for applications where the BPC contents are post filtered.

Product Features:
• Mixtainer BioProcess Container — supplied sterilized by irradiation and is constructed of Thermo Scientific CX5-14 film and contains a magnetic stir-bar located in a plastic dish welded in the bottom of the BPC. The dish contains a locking ring to ensure the containment of the stir-bar during shipping, handling and operation. A powder port option is available on one of the standard configurations. Aseptic liquid transfers are facilitated by connectors and tubing attached to ports at the top of the BPC


Recommended for:
• Hydration of powdered media and buffers
• Sterile mixing of cell culture media with sera and reagents
• Sterile mixing of buffer solutions or other process fluids
• Pooling of sterile liquid fractions

Specifications
DescriptionMixtainer BioProcess Container, 50 L
Film DescriptionCX5-14
Outer PackagingPackages placed in strong cardboard cartons with product and lot identification
Port DetailPowder addition port 38.1 mm (1.5 in.) tri-clamp port, Top Ports: 2 x 0.25 in. 2 x 0.38 in.
Product LineHyPerforma, Mixtainer
Protective Inner PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity50 L
Sterilization MethodGamma Irradiated
Tubing Line 1Line 1. 9.5 mm (0.38 in.) Quick Connect Body with 16.5 cm (6.5 in.) dip tube, tube length: 91 cm (36 in.)
Tubing Line 2Line 2. 6.3 mm (0.25 in.) Insert luer lock, tube length: 91 cm (36 in.)
Tubing Line 3Line 3. 6.3 mm (0.25 in.) Insert luer lock, tube length: 91 cm (36 in.)
Tubing Line 4Line 4. 9.53 mm (0.38 in.) Quick Connect, body tube length: 91 cm (36 in.)
Capacity (Metric)50 L
Unit Size50 L