Labtainer™ BioProcess Container (BPC), 50L with 3 Ports, MPC Insert, 2D BPC
Thermo Scientific™

Labtainer™ BioProcess Container (BPC), 50L with 3 Ports, MPC Insert, 2D BPC

The 50 L Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single web, multi-layer plastic film and includes자세히 알아보기
Have Questions?
카탈로그 번호수량
SH3071404
SH30714.04으로도 사용됨
50 L, each
카탈로그 번호 SH3071404
SH30714.04으로도 사용됨
제품 가격(KRW)
-
수량:
50 L, each
The 50 L Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single web, multi-layer plastic film and includes 3 top ports (2 with MPC inserts, 1 plugged port).

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All Labtainers are constructed in an ISO 7 cleanroom under cGMP conditions.

Applications of the Thermo Scientific Labtainer BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors.
• Buffer, culture media, and process liquid preparation and storage.
• Chromatography feed and fraction collection.
• Harvest, storage and transport of bulk drug product and bulk drug precursors.

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library – with over 20,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
Filters—used for bioburden reduction; sterilization grade filters and vent filters.
Tubing—thermoplastic elastomers, platinum-cured silicone.
Sample ports—septum-style and needle-free ports.
Clamps—sliders, pinch, and crimp bands.

사양
치수82.6 cm x 58.4 cm (32.5 in. x 23 in.)
외부 포장Packages placed in strong cardboard cartons with product and lot identification
제품라인Labtainer
보호용 내부 포장Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
수량50 L, each
멸균 방법Gamma Irradiated
튜빙 라인 1Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 30 cm (12 in.) ID x OD: 9.5 x 12.7 mm (0.38 x 0.5 in.)
튜빙 라인 2Line 2. MPC insert, polycarbonate, Tubing: C-Flex, length: 30 cm (12 in.) ID x OD: 9.5 x 12.7mm (0.38 x 0.5 in.)
튜빙 라인 3Line 3. Plugged, No Tubing
용량(미터법)50 L
Film DescriptionCX5-14
Port DetailEdge Ports: 2 x 0.38 in. 1 x 0.25 in.
Unit Size50 L