PowderTainer™ II 1 to 25kg Single-Use BPC for powder applications, 25kg
Thermo Scientific™

PowderTainer™ II 1 to 25kg Single-Use BPC for powder applications, 25kg

The 25 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 filmRead more
Have Questions?
Catalog NumberCapacity (Metric)
SH3073703
also known as SH30737.03
25 kg
Catalog number SH3073703
also known as SH30737.03
Price (MXN)
-
Capacity (Metric):
25 kg
The 25 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 film that includes 2 ports (1 wash down line, 1 powder port). The Powdertainer BPC is specifically designed for powder containment and discharge applications, and maintains a closed system for maximum recovery of powder while minimizing the risk of cross contamination.

Features:
• Three-inch tri-clamp port designed for secure connection to, and easy integration, with existing formulation vessels.
• Powdertainer BPC features suspensor handle for support during discharge, as well as neck clamp to retain powder prior to discharge.
• Use this system to eliminate post-use cleaning steps required with re-useable containers.

Applications of the Thermo Scientific Powdertainer BPC include:
• Storage and delivery of powder culture media and buffers.
• Storage and dispensing of pre-weighed chemicals and other process powders.

Specifications
Capacity (English)6.6 gal.
Capacity (Metric)25 kg
DescriptionDisposable cell culture/bioprocess bag for powder applications
Dimensions (L x W x H)83.3cm x 59.2cm (32.8 in. x 23.3 in.)
Film DescriptionCX3-9
Includes2 Ports (1 Wash Down Line, 1 Powder Port)
Port DetailPowder port: 7.6cm (3 in.) tri-clamp with integrated seal
Product LinePowderTainer™ II
Washdown LineYes
DisposableSingle-use
Product TypeBioprocess Container for Powder