Product Image
Impeller sleeve for open-top mixing, for 50L HyPerforma™ Single-Use Mixer BioProcess Container (BPC)
Have Questions?
Número de catálogo SH3074906
también denominado SH30749.06
Precio (EUR)
-
The Impeller sleeve for open-top mixing (50L) supports mixing applications in bioproduction using Thermo Scientific™ Bioprocess Containers (BPCs) for Thermo Scientific™ HyPerforma™ Single-Use Mixer, with impeller-based mixing in open or closed designs.
Product documentation:
To search for Certificates of Analysis (COA) and other product certificates and documentation by part number, lot number or catalog number, click here.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability