Thermo Scientific™
BioProcess Containers (BPC), 200, for HyPerforma™ Single-Use Mixer and docking tank, liquid-to-liquid w/probe ports This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.
Catalog number:  SH30753.02
Related applications:
Bioprocessing Containers
Back to top
Description
The 200L Thermo Scientific™ Bioprocess Container (BPC) with probe ports is for use with the Thermo Scientific™ HyPerforma™ Single-Use Mixer and docking tank for liquid-to-liquid mixing.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Specifications
Green Features: Fewer resources used and less waste
Product Line: HyPerforma™
Product Size: 200 L