Product Image
BioProcess Containers (BPC), 500L, for HyPerforma™ Single-Use Mixer and docking tank, liquid-to-liquid w/probe ports
Have Questions?
Número de catálogo SH3075402
Precio (MXN)
-
The 500L Thermo Scientific™ Bioprocess Container (BPC) with probe ports is for use with the Thermo Scientific™ HyPerforma™ Single-Use Mixer and docking tank for liquid-to-liquid mixing.
Product documentation:
To search for Certificates of Analysis (COA) and other product certificates and documentation by part number, lot number or catalog number, click here.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Especificaciones
Unit Size500 L