Thermo Scientific™
BioProcess Containers (BPC), 1000L, for HyPerforma™ Single-Use Mixer and docking tank, liquid-to-liquid w/probe ports
Catalog number: SH30755.02
Thermo Scientific™
BioProcess Containers (BPC), 1000L, for HyPerforma™ Single-Use Mixer and docking tank, liquid-to-liquid w/probe ports
Catalog number: SH30755.02
Catalog Number
SH30755.02
Unit Size
Price (USD)
Catalog NumberUnit SizePrice (USD)
SH30755.02Request A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The 1000L Thermo Scientific™ Bioprocess Container (BPC) with probe ports is for use with the Thermo Scientific™ HyPerforma™ Single-Use Mixer and docking tank for liquid-to-liquid mixing.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Specifications
Capacity (Metric)
1000 L
Film Description
CX5-14
Port Detail
3 Probe Port
For Use With (Application)
Liquid-Liquid BPC
Product Line
HyPerforma™
Green Features
Fewer resources used and less waste