The 50L standard, open-top tank liner, constructed of CX3-9 film, is for use with the Thermo Scientific™ HyPerforma™ Single-Use Mixer and docking tank for mixing applications.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability