Thermo Scientific™

BioProcess Container (BPC), 2000L, for HyPerforma™ Single-Use Mixer, open-top mixing liner This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  SH30762.05

 Related applications:

Bioprocessing Containers

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The 2000L standard, open-top tank liner, constructed of CX3-9 film, is for use with the Thermo Scientific™ HyPerforma™ Single-Use Mixer and docking tank for mixing applications.

Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability

Specifications

Film Description: CX3-9
Green Features: Fewer resources used and less waste
Port Detail: 0 Probe Port
Product Line: HyPerforma™
Size (Metric): 2,000 L

Documents

Certificates of Analysis

Search for Certificates of Analysis for your custom and standard Thermo Scientific™ single-use products.

Manuals & protocols