BioProcess Container (BPC), 2000L, for HyPerforma™ Single-Use Mixer, liquid-to-liquid w/probe ports
Thermo Scientific™
BioProcess Container (BPC), 2000L, for HyPerforma™ Single-Use Mixer, liquid-to-liquid w/probe ports
Have Questions?
Número de catálogoCapacidad (métrico)
SH30769.022000 L
Número de catálogo SH30769.02
Precio (MXN)
-
Capacidad (métrico):
2000 L
The 2000L Thermo Scientific™ Bioprocess Container (BPC) for liquid-to-liquid mixing applications, is for use with the Thermo Scientific™ HyPerforma™ Single-Use Mixer and is composed of CX 5-14 single web, multi-layer plastic film and includes probe ports.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Especificaciones
Descripción de la películaCX5-14
Para utilizar con (aplicación)Liquid-Liquid BPC
Detalle del puerto3 Probe Port
Línea de productosHyPerforma
Capacidad (métrico)2000 L