Product Image
Thermo Scientific™
Sampling manifold accessory for HyPerforma™ Single-Use Bioreactors, Single pack - includes Luer Locks
Have Questions?
Número de catálogoCantidad
SH30845.011 colector/envase individual
Número de catálogo SH30845.01
Precio (MXN)
-
Cantidad:
1 colector/envase individual
The Sterile Sampling Manifold with Luer Locks (single-pack) is an accessory for the Thermo Scientific™ HyPerforma™ Single-Use Bioreactors and is available to weld onto the sample line to take sample sets.
Especificaciones
Línea de productosHyPerforma
Cantidad1 colector/envase individual
Para utilizar con (equipo)Single-Use Bioreactor
Unit Size1 manifold⁄single-pack