PowderTainer™ II 1 to 25kg Single-Use BPC for powder applications, 1 kg
Thermo Scientific™

PowderTainer™ II 1 to 25kg Single-Use BPC for powder applications, 1 kg

The 1 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 filmMás información
Have Questions?
Número de catálogoCapacidad (métrico)
SH3086401
también denominado SH30864.01
1 kg
Número de catálogo SH3086401
también denominado SH30864.01
Precio (MXN)
-
Capacidad (métrico):
1 kg
The 1 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 film that includes 1 port (1 powder port). The Powdertainer BPC is specifically designed for powder containment and discharge applications, and maintains a closed system for maximum recovery of powder while minimizing the risk of cross contamination.

Features:
• Three-inch tri-clamp port designed for secure connection to, and easy integration, with existing formulation vessels.
• Powdertainer BPC features suspensor handle for support during discharge, as well as neck clamp to retain powder prior to discharge.
• Use this system to eliminate post-use cleaning steps required with re-useable containers.

Applications of the Thermo Scientific Powdertainer BPC include:
• Storage and delivery of powder culture media and buffers.
• Storage and dispensing of pre-weighed chemicals and other process powders.

Especificaciones
Capacidad (métrico)1 kg
Descripción de la películaCX3-9
Incluye1 Port (1 Powder Port)
Longitud (métrico)42.7 cm
Embalaje exteriorLos paquetes se colocan en cajas de cartón resistentes con identificación de productos y lotes.
Detalle del puertoPuerto de polvo: 3 in
Línea de productosPowderTainer™ II
Embalaje interior de protecciónEnvasado y sellado en dos capas exteriores independientes en el área de clase 10 000/ISO clase 7
Método de esterilizaciónSin irradiación
Línea de tuberías 1Línea de lavado: N/D
Línea de tuberías 2Powder port: 7.6 cm (3 in.) tri-clamp with intergrated seal
Anchura (métrico)31.8 cm
Tipo de productoBioprocess Container for Powder