3D Tank Liner, Aegis™ 5-14 film, top and bottom drain
Thermo Scientific™
3D Tank Liner, Aegis™ 5-14 film, top and bottom drain
Thermo Scientific 3D Tank Liners are composed of Aegis 5-14 film with dimensions optimized for Thermo Scientific drums and commonly-used,Más información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)ForUseWithApplication
SH3098802100 L
SH309880150 L
SH3098803200 L
Número de catálogo SH3098802
Precio (MXN)
-
Capacidad (métrico):
100 L
Thermo Scientific 3D Tank Liners are composed of Aegis 5-14 film with dimensions optimized for Thermo Scientific drums and commonly-used, industry-standard cylindrical tanks. The tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
• Minimizes bioburden due to construction under sterile cGMP conditions
• Helps eliminate post-use cleaning steps required with reusable containers, reducing cross-contamination risks and reduce cycle times
• Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Especificaciones
Cantidad100 L
Capacidad (métrico)100 L
Tubing StyleC-Flex
Unit Size100 L