Labtainer™ 2D BioProcess Container (BPC), three ports, MPCs, quick-connect
Labtainer™ 2D BioProcess Container (BPC), three ports, MPCs, quick-connect
Thermo Scientific™
Labtainer™ 2D BioProcess Container (BPC), three ports, MPCs, quick-connect
Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis5-14 single-web, multi-layer film.Features of the Labtainer 2D BPC include:•Más información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)
SH31005015 L
SH310050210 L
SH310050320 L
SH310050450 L
SH31005052 L
Número de catálogo SH3100501
Precio (MXN)
-
Capacidad (métrico):
5 L
Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis5-14 single-web, multi-layer film.
Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions
Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
Especificaciones
Capacidad (métrico)5 L
Certificaciones/conformidadISO 7
Descripción de la películaAegis 5-14
Para utilizar con (aplicación)Bioprocessing
Detalle del puerto3 ports
Cantidad5 L
Estilo de los tubosC-Flex
Esterilizables en autoclaveNot Autoclavable
ColorTransparente
DesechablesSingle-use
EsterilidadEstéril
Unit Size5 L