Thermo Scientific™ Single-Use Fermentor BioProcess Container, Aegis™ 5-14 film, pH/DO sensor, foam sensor, Meissner inlet, two 10-inch exhaust filters, 300 L
Thermo Scientific™
Thermo Scientific™ Single-Use Fermentor BioProcess Container, Aegis™ 5-14 film, pH/DO sensor, foam sensor, Meissner inlet, two 10-inch exhaust filters, 300 L
This 300 L Thermo Scientific Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of Aegis 5-14 film, a five-layer, animal-originMás información
Have Questions?
Número de catálogoCantidad
SH3100902300 L
Número de catálogo SH3100902
Precio (MXN)
-
Cantidad:
300 L
This 300 L Thermo Scientific Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of Aegis 5-14 film, a five-layer, animal-origin free film, and includes a Mettler Toledo single-use pH and DO sensor, foam sensor, Meissner inlet, two 10-inch exhaust filters, and condenser.
All SUF BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
Features:
• Manufactured with industry-leading Aegis 5-14 film
• C-flex tubing that can be used with tubing welder and various connection options
• Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
Filters—used for bioburden reduction; sterilization grade filters and gas filters
Tubing—thermoplastic elastomers, platinum-cured silicone
Sample ports—septum-style and needle-free ports
Clamps—sliders, pinch, and crimp bands
Especificaciones
Línea de productosHyPerforma
Cantidad300 L
Capacidad (métrico)300 L
Unit Size300 L
Contenido y almacenamiento
Store at room temperature.