Thermo Scientific™

Thermo Scientific™ Single-Use Fermentor BioProcess Container, CX5-14 film, pH/DO sensor, foam sensor, Meissner inlet, one 5-inch exhaust filter, 30 L

Catalog number:  SH3101901

 Related applications:

Bioprocessing Containers

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This 30 L Thermo Scientific Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of CX5-14 film, a five-layer, animal-origin free film, and includes a Mettler Toledo single-use pH and DO sensor, foam sensor, Meissner inlet, one 5-inch exhaust filter, and condenser.

All SUF BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.

Features:
• Manufactured with industry-leading CX5-14 film
• C-flex tubing that can be used with tubing welder and various connection options
• Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
Filters—used for bioburden reduction; sterilization grade filters and gas filters
Tubing—thermoplastic elastomers, platinum-cured silicone
Sample ports—septum-style and needle-free ports
Clamps—sliders, pinch, and crimp bands

Specifications

Product Line: HyPerforma™
Product Size: 30 L
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols