Thermo Scientific™ Single-Use Fermentor BioProcess Container, CX5-14 film, traditional ports, foam sensor, Meissner inlet, two 10-inch exhaust filters, 300 L
Thermo Scientific™
Thermo Scientific™ Single-Use Fermentor BioProcess Container, CX5-14 film, traditional ports, foam sensor, Meissner inlet, two 10-inch exhaust filters, 300 L
This 300 L Thermo Scientific Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of CX5-14 film, a five-layer, animal-origin free詳細を見る
製品番号(カタログ番号)数量
SH3103005300 L
製品番号(カタログ番号) SH3103005
価格(JPY)
-
数量:
300 L
This 300 L Thermo Scientific Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of CX5-14 film, a five-layer, animal-origin free film, and includes traditional ports, foam sensor, Meissner inlet, two 10-inch exhaust filters, and condenser.
All SUF BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
Features:
• Manufactured with industry-leading CX5-14 film
• C-flex tubing that can be used with tubing welder and various connection options
• Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
Filters—used for bioburden reduction; sterilization grade filters and gas filters
Tubing—thermoplastic elastomers, platinum-cured silicone
Sample ports—septum-style and needle-free ports
Clamps—sliders, pinch, and crimp bands
仕様
製品ラインHyPerforma
数量300 L
容量(メートル法)300 L
Unit Size300 L
組成および保存条件
Store at room temperature.