BioProcess Container (BPC), 100L, for HyPerforma™ Single-Use Mixer, powder-to-liquid, CX5-14 film, 0 ports
Thermo Scientific™
BioProcess Container (BPC), 100L, for HyPerforma™ Single-Use Mixer, powder-to-liquid, CX5-14 film, 0 ports
The 100L Thermo Scientific™ Bioprocess Containers (BPC) for powder-to-liquid mixing, is composed of CX5-14 single web, multi-layer plastic film andMás información
Have Questions?
Número de catálogoQuantity
SH31046.01
Número de catálogo SH31046.01
Precio (MXN)
-
The 100L Thermo Scientific™ Bioprocess Containers (BPC) for powder-to-liquid mixing, is composed of CX5-14 single web, multi-layer plastic film and includes 0 ports.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Especificaciones
Descripción de la películaCX5-14
Para utilizar con (aplicación)Powder-Liquid BPC
Detalle del puerto0 Probe Port
Línea de productosHyPerforma
Capacidad (métrico)100 L