BioProcess Container (BPC), 200L, for HyPerforma™ Single-Use Mixer, powder-to-liquid, Aegis™ 5-14 film, 3 ports
Thermo Scientific™
BioProcess Container (BPC), 200L, for HyPerforma™ Single-Use Mixer, powder-to-liquid, Aegis™ 5-14 film, 3 ports
The 200L Thermo Scientific™ Bioprocess Containers (BPC) for powder-to-liquid mixing, is composed of Aegis 5-14 single web, multi-layer plastic filmMás información
Have Questions?
Número de catálogoCantidad
SH31053.02Cada uno
Número de catálogo SH31053.02
Precio (MXN)
-
Cantidad:
Cada uno
The 200L Thermo Scientific™ Bioprocess Containers (BPC) for powder-to-liquid mixing, is composed of Aegis 5-14 single web, multi-layer plastic film and includes 3 ports.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Especificaciones
Detalle del puerto3 ports
Línea de productosHyPerforma
CantidadCada uno
Capacidad (métrico)200 L
Unit Size200 L