BioProcess Container (BPC), 200L, for HyPerforma™ Single-Use Mixer, liquid-to-liquid, CX5-14 film, 3 ports
Thermo Scientific™
BioProcess Container (BPC), 200L, for HyPerforma™ Single-Use Mixer, liquid-to-liquid, CX5-14 film, 3 ports
The 200L Thermo Scientific™ Bioprocess Containers (BPC) for liquid-to-liquid mixing, is composed of CX5-14 single web, multi-layer plastic film andRead more
Have Questions?
Catalog NumberQuantity
SH31054.04
also known as SH3105404
Catalog number SH31054.04
also known as SH3105404
Price (MXN)
-
The 200L Thermo Scientific™ Bioprocess Containers (BPC) for liquid-to-liquid mixing, is composed of CX5-14 single web, multi-layer plastic film and includes 3 ports.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Specifications
Film DescriptionCX5-14
For Use With (Application)Liquid-Liquid BPC
Port Detail3 Probe Port
Product LineHyPerforma
Capacity (Metric)200 L