This 2000 L Thermo Scientific 5:1 Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of Aegis 5-14 film, a five-layer,자세히 알아보기
Have Questions?
카탈로그 번호
수량
SH3113601
2,000 L
카탈로그 번호 SH3113601
제품 가격(KRW)
-
수량:
2,000 L
This 2000 L Thermo Scientific 5:1 Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of Aegis 5-14 film, a five-layer, animal-origin free film, and includes a drilled-hole sparger (DHS), crossflow sparger, and condenser.
All SUF BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
Features: • Manufactured with industry-leading Aegis 5-14 film • C-flex tubing that can be used with tubing welder and various connection options • Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology Choose from the industry’s largest component library, with over 2,000 unique components that include: • Fittings—straight, reducing, elbow, T-style, X-style, and cross-style • Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors • Filters—used for bioburden reduction; sterilization grade filters and gas filters • Tubing—thermoplastic elastomers, platinum-cured silicone • Sample ports—septum-style and needle-free ports • Clamps—sliders, pinch, and crimp bands