HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
Thermo Scientific™
HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
This bioprocess container (BPC) is for use with the Thermo Scientific HyPerforma Smartainer 3.0, which provides end-users producing biologics or자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)인서트 유형
SH31188.01200 LHorizontally-gusseted
SH31188.02500 LHorizontally-gusseted
SH31188.03500 LVertically-gusseted
SH31188.041000 LVertically-gusseted
SH31188.051500 LVertically-gusseted
SH31188.062000 LVertically-gusseted
SH31188.072500 LVertically-gusseted
SH31188.083000 LVertically-gusseted
카탈로그 번호 SH31188.01
제품 가격(KRW)
-
용량(미터법):
200 L
인서트 유형:
Horizontally-gusseted
This bioprocess container (BPC) is for use with the Thermo Scientific HyPerforma Smartainer 3.0, which provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L. The Smartainer 3.0 system is an update to the Smartainer 2.0 system, and is available in either non-jacketed or jacketed configuation. The Smartainer 3.0 system is designed for the collection and storage of sterile process liquids such as cell culture media, process water, or purification media—wherever large volumes of sterile process liquids need to be safely, securely stored.
BPCs to optimize performance
Choose from a variety of off-the-shelf BPCs or customize a BPC to fit your Smartainer 3.0 system. Customization options include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
사양
인서트 유형Horizontally-gusseted
용량(미터법)200 L
Product TypeBioprocess Container (BPC)
Unit Size200 L