HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
Thermo Scientific™
HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
This bioprocess container (BPC) is for use with the Thermo Scientific HyPerforma Smartainer 3.0, which provides end-users producing biologics orMás información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)Tipo de inserto
SH31188.041000 LCon fuelle vertical
SH31188.01200 LCon fuelle horizontal
SH31188.02500 LCon fuelle horizontal
SH31188.03500 LCon fuelle vertical
SH31188.051500 LCon fuelle vertical
SH31188.062000 LCon fuelle vertical
SH31188.072500 LCon fuelle vertical
SH31188.083000 LCon fuelle vertical
Número de catálogo SH31188.04
Precio (USD)
-
Capacidad (métrico):
1000 L
Tipo de inserto:
Con fuelle vertical
This bioprocess container (BPC) is for use with the Thermo Scientific HyPerforma Smartainer 3.0, which provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L. The Smartainer 3.0 system is an update to the Smartainer 2.0 system, and is available in either non-jacketed or jacketed configuation. The Smartainer 3.0 system is designed for the collection and storage of sterile process liquids such as cell culture media, process water, or purification media—wherever large volumes of sterile process liquids need to be safely, securely stored.
BPCs to optimize performance
Choose from a variety of off-the-shelf BPCs or customize a BPC to fit your Smartainer 3.0 system. Customization options include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
Especificaciones
Tipo de insertoCon fuelle vertical
Capacidad (métrico)1000 L
Product TypeBioprocess Container (BPC)
Unit Size1000 L