HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
Thermo Scientific™
HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
This bioprocess container (BPC) is for use with the Thermo Scientific HyPerforma Smartainer 3.0, which provides end-users producing biologics orLeia mais
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoCapacity (Metric)Insert Type
SH31188.051500 LVertically-gusseted
SH31188.01200 LHorizontally-gusseted
SH31188.02500 LHorizontally-gusseted
SH31188.03500 LVertically-gusseted
SH31188.041000 LVertically-gusseted
SH31188.062000 LVertically-gusseted
SH31188.072500 LVertically-gusseted
SH31188.083000 LVertically-gusseted
Número do catálogo SH31188.05
Preço (BRL)
-
Capacity (Metric):
1500 L
Insert Type:
Vertically-gusseted
This bioprocess container (BPC) is for use with the Thermo Scientific HyPerforma Smartainer 3.0, which provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L. The Smartainer 3.0 system is an update to the Smartainer 2.0 system, and is available in either non-jacketed or jacketed configuation. The Smartainer 3.0 system is designed for the collection and storage of sterile process liquids such as cell culture media, process water, or purification media—wherever large volumes of sterile process liquids need to be safely, securely stored.
BPCs to optimize performance
Choose from a variety of off-the-shelf BPCs or customize a BPC to fit your Smartainer 3.0 system. Customization options include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
Especificações
Insert TypeVertically-gusseted
Capacity (Metric)1500 L
Product TypeBioprocess Container (BPC)
Unit Size1500 L