Thermo Scientific™
Harvestainer™ Microcarrier Separation System
Catalog number: SH31267.02
Thermo Scientific™
Harvestainer™ Microcarrier Separation System
Catalog number: SH31267.02
Catalog Number
SH31267.02
Unit Size
50 L
Price (USD)
Catalog Number
Unit Size
25 L
Price (USD)
Catalog NumberUnit SizePrice (USD)
SH31267.0250 L
SH31267.0125 L
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
The Thermo Scientific Harvestainer Microcarrier Separation System is the ideal solution for the separation of microcarrier beads during adherent cell culture production. The Harvestainer system is a closed, single-use system that helps to increase product yields versus traditional methods, while reducing clean-in-place and steam-in-place requirements and water for injection usage. It eliminates equipment maintenance and reduces process validations, cycle times, and manual final decanting processes.
For smaller-scale or larger-scale applications, the Harvestainer system is ideal for in-line microcarrier separation. The 3 L and 12 L Harvestainer systems consist of a pre-assembled bioprocess container (BPC) and tray in a single-use unit, where the tray acts as the secondary containment device. The 25 L and 50 L Harvestainer systems consist of a 3D BPC with a drum as the secondary containment device. The BPC features a dip tube with dipwell design for better drainage and minimal manipulation.
• Closed system
• Scalable for both smaller-scale and larger-scale applications
• Helps deliver high recovery rates
• Inline microcarrier separation
Applications
• Adherent cell culture production
Specifications
Tubing Style
C-Flex
Volume (Metric)
50 L
For Use With
Single-Use Bioprocessing
Figures