Labtainer™ 2D BioProcess Container, BioTitan retainers, three ports, Luer locks
Thermo Scientific™

Labtainer™ 2D BioProcess Container, BioTitan retainers, three ports, Luer locks

The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film. Features of the ThermoRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SH31294.052 L
SH31294.015 L
SH31294.0210 L
SH31294.0320 L
SH31294.0450 L
Catalog number SH31294.05
Price (EUR)
-
Capacity (Metric):
2 L

The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.

Features of the Thermo Scientific 2D BPC system include:
• Now manufactured with the BioTitan Retention Device—designed to provide superior universal retention that minimizes the risk of leakage and failure at the connection point
• Use to eliminate post-use cleaning steps required with re-useable containers and to reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions

Applications of the Thermo Scientific 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Buffer, culture media, and process liquid preparation and storage
• Chromatography feed and fraction collection
• Harvest, storage, and transport of bulk drug product and bulk drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Specifications
Quantity2 L
Capacity (Metric)2 L
Tubing StyleTPE
Unit Size2 L