Transfer Assembly Pump Tubing
Thermo Scientific™
Transfer Assembly Pump Tubing
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. Simplified connectivity and greater process efficiency for most upstream and downstream process operations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesTubing Style
SH31320.01MPC Insert and MPC Body3/8 in. TPE and Pumpsil Silicone
SH31314.01MPU Insert and MPU Body3/4 in. TPE and Pumpsil Silicone
SH31406.01AseptiQuik G Connectors1/4 in. ID Silicone and Pumpsil
SH31406.11AseptiQuik G Connectors3/8 in. ID Silicone and Pumpsil
SH31406.21AseptiQuik G Connectors1/2 in. ID Silicone and Pumpsil
SH31406.31AseptiQuik G Connectors3/4 in. ID Silicone and Pumpsil
SH31407.01AseptiQuik L Connectors3/4 in. ID Silicone/Pumpsil Silicone
SH31407.11AseptiQuik L Connectors1 in. ID Silicone and Pumpsil
SH31408.01MPC Insert and MPC Body3/8 in. ID Silicone and Pumpsil
SH31409.01MPU Insert and MPU Body3/4 in. ID Silicone and Pumpsil
SH31410.01MPU Insert and MPU Body1 in. ID Silicone and Pumpsil
SH31318.01MPX insert and MPX body1/2 in. TPE and Pumpsil Silicone
SH31411.01MPX Insert and MPX Body1/2 in. ID Silicone and Pumpsil
SH31412.01AseptiQuik G Connectors1/4 in. ID Braided Silicone, Pumpsil
SH31333.01MPU Insert and MPU Body1 in. TPE and Pumpsil Silicone
SH31412.11AseptiQuik G Connectors3/8 in. ID Braided Silicone, Pumpsil
SH31341.01AseptiQuik L Connectors3/4 in. TPE and Pumpsil Silicone
SH31412.21AseptiQuik G Connectors1/2 in. ID Braided Silicone, Pumpsil
SH31341.11AseptiQuik L Connectors1 in. TPE and Pumpsil Silicone
SH31412.31AseptiQuik G Connectors3/4 in. ID Braided Silicone, Pumpsil
SH31342.01AseptiQuik G Connectors1/4 in. TPE and Pumpsil Silicone
SH31413.01AseptiQuik L Connectors3/4 in. ID Braided Silicone, Pumpsil
SH31342.11AseptiQuik G Connectors3/8 in. TPE and Pumpsil Silicone
SH31413.11AseptiQuik L Connectors1 in. ID Braided Silicone, Pumpsil
SH31342.21AseptiQuik G Connectors1/2 in.TPE and Pumpsil Silicone
SH31414.01MPC Insert and MPC Body3/8 in. ID Braided Silicone, Pumpsil
SH31342.31AseptiQuik G Connectors3/4 in.TPE and Pumpsil Silicone
SH31415.01MPU Insert and MPU Body3/4 in. ID Braided Silicone, Pumpsil
SH31416.01MPU Insert and MPU Body1 in. ID Braided Silicone, Pumpsil
SH31417.01MPX Insert and MPX Body1/2 in. ID Braided Silicone, Pumpsil
Catalog number SH31320.01
Price (MXN)
-
Connector Types:
MPC Insert and MPC Body
Tubing Style:
3/8 in. TPE and Pumpsil Silicone
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.
These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.
Specifications
Connector TypesMPC Insert and MPC Body
DescriptionTransfer Assembly Pump Tubing, 3/8 inch ID TPE, 3/8 inch ID Pumpsil Silicone, MPC Insert and MPC Body
QuantityEach
Tubing MaterialPharmaceutical-grade thermoplastic elastomer, platinum-cured silicone
Tubing Style3/8 in. TPE and Pumpsil Silicone
Unit Size1 each
Contents & Storage
Room Temperature