HyPerforma™ Single-Use Mixer BioProcess Container, Aegis™ 5-14 film
HyPerforma™ Single-Use Mixer BioProcess Container, Aegis™ 5-14 film
Thermo Scientific™
HyPerforma™ Single-Use Mixer BioProcess Container, Aegis™ 5-14 film
The Thermo Scientific™ HyPerforma™ Single-Use Mixer BioProcess Container (BPC) aids in providing greater process efficiency for most upstream and downstream process operations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Port Detail
SH31345.01100 LWith Ports
SH31346.01100 LWithout Ports
SH31347.01200 LWith Ports
SH31348.01200 LWithout Ports
SH31349.01500 LWith Ports
SH31350.01500 LWithout Ports
SH31351.011000 LWith Ports
SH31352.011000 LWithout Ports
SH31353.012000 LWith Ports
SH31354.012000 LWithout Ports
Catalog number SH31345.01
Price (MXN)
-
Request A Quote
Capacity (Metric):
100 L
Port Detail:
With Ports
The Thermo Scientific™ HyPerforma™ Single-Use Mixer BioProcess Container (BPC) is designed for use with the HyPerforma Single-Use Mixer, and is used for both upstream and downstream mixing applications. Together, these products help deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
The Thermo Scientific™ HyPerforma™ Single-Use Mixer BioProcess Container (BPC) is designed for use with the HyPerforma Single-Use Mixer, and is used for both upstream and downstream mixing applications. Together they deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations. The complete line of HyPerforma Single-Use Mixers include 50, 100, 200, 500, 1000 and 2000 L sizes, helping ensure consistent scalability for a number of applications from powder media and buffer hydration to advanced downstream harvest and intermediate pooling steps. Together, these products help deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Thermo Scientific delivers a comprehensive and complete set of BPCs and other single-use products to meet the needs of your facility or process—designed to aid many common production processes such as:
• Powder media hydration
• Liquid media concentrate dilution
• Harvest hold tank
• Viral inactivation
• Downstream intermediate pooling
• Final product hold and additive suspension
Specifications
Capacity (Metric)100 L
Description100 L HyPerforma Single-Use Mixer (S.U.M.) BioProcess Container (BPC), Aegis 5-14 film with ports
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
Port DetailWith Ports
QuantityEach
Unit Size1 each
Contents & Storage
Room Temperature