Search
Search

| Número de catálogo | Capacidad (métrico) | Detalle del puerto |
|---|---|---|
| SH31349.01 | 500 L | Con puertos |
| SH31345.01 | 100 L | Con puertos |
| SH31346.01 | 100 L | Sin puertos |
| SH31347.01 | 200 L | Con puertos |
| SH31348.01 | 200 L | Sin puertos |
| SH31350.01 | 500 L | Sin puertos |
| SH31351.01 | 1000 L | Con puertos |
| SH31352.01 | 1000 L | Sin puertos |
| SH31353.01 | 2000 L | Con puertos |
| SH31354.01 | 2000 L | Sin puertos |
The Thermo Scientific™ HyPerforma™ Single-Use Mixer BioProcess Container (BPC) is designed for use with the HyPerforma Single-Use Mixer, and is used for both upstream and downstream mixing applications. Together they deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations. The complete line of HyPerforma Single-Use Mixers include 50, 100, 200, 500, 1000 and 2000 L sizes, helping ensure consistent scalability for a number of applications from powder media and buffer hydration to advanced downstream harvest and intermediate pooling steps. Together, these products help deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Thermo Scientific delivers a comprehensive and complete set of BPCs and other single-use products to meet the needs of your facility or process—designed to aid many common production processes such as:
• Powder media hydration
• Liquid media concentrate dilution
• Harvest hold tank
• Viral inactivation
• Downstream intermediate pooling
• Final product hold and additive suspension