imPULSE™ Single-Use Mixer and BioProcess Containers, ASI™ 26/77 Film
imPULSE™ Single-Use Mixer and BioProcess Containers, ASI™ 26/77 Film
imPULSE™ Single-Use Mixer and BioProcess Containers, ASI™ 26/77 Film
Thermo Scientific™
imPULSE™ Single-Use Mixer and BioProcess Containers, ASI™ 26/77 Film
The Thermo Scientific imPULSE Single-Use Mixer BioProcess Container (BPC) aids in providing greater process efficiency for most upstream and downstream process operations.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)Detalle del puerto
SH31359250 LSin puertos
SH31358100 LCon puertos
SH31357100 LSin puertos
SH31360250 LCon puertos
SH31362500 LCon puertos
SH31361500 LSin puertos
SH313641000 LCon puertos
SH313631000 LSin puertos
SH313662000 LCon puertos
SH313652000 LSin puertos
SH313683000 LCon puertos
SH313673000 LSin puertos
SH313705000 LCon puertos
SH313695000 LSin puertos
Número de catálogo SH31359
Precio (MXN)
-
Capacidad (métrico):
250 L
Detalle del puerto:
Sin puertos
The Thermo Scientific imPULSE Single-Use Mixer BioProcess Container (BPC) is designed for use with the imPULSE Single-Use Mixer, and is used for both upstream and downstream mixing applications. Together, these products help deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
  • Designed for use with the imPULSE Single-Use Mixer
  • Used for both upstream and downstream mixing applications
  • Together, these products help deliver simplified connectivity and enable greater process efficiency
Especificaciones
Descripción250 L imPULSE Single-Use Mixer (S.U.M.) BioProcess Container (BPC), ASI 26/77 film without ports
Descripción de la películaASI 26/77 Film
Para utilizar con (aplicación)Monoclonal antibody production, upstream and downstream applications
Detalle del puertoSin puertos
CantidadEach
Capacidad (métrico)250 L
Unit Size250 L
Contenido y almacenamiento
Ambient/room temperature