| SH31407.11 | Full specifications | 1 each | Transfer Assembly Pump Tubing, 1 in. ID Silicone and Pumpsil Silicone, AseptiQuik L Connectors | Request A Quote |
Description | Transfer Assembly Pump Tubing, 1 in. ID Silicone and Pumpsil Silicone, AseptiQuik L Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31314.01 | Full specifications | 1 each | Transfer Assembly Pump Tubing, 3/4 inch ID TPE and Pumpsil Silicone, MPU Insert and MPU Body | Request A Quote |
Description | Transfer Assembly Pump Tubing, 3/4 inch ID TPE and Pumpsil Silicone, MPU Insert and MPU Body | Quantity | Each | Tubing Style | TPE and Pumpsil Silicone | Connector Types | MPU Insert and MPU Body | Tubing Material | Pharmaceutical-grade thermoplastic elastomer, platinum-cured silicone | Unit Size | 1 each |
|
| SH31406.01 | Full specifications | 1 each | Transfer Assembly Pump Tubing, 1/4 in. ID Silicone and Pumpsil Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly Pump Tubing, 1/4 in. ID Silicone and Pumpsil Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31406.11 | Full specifications | 1 each | Transfer Assembly Pump Tubing, 3/8 in. ID Silicone and Pumpsil Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly Pump Tubing, 3/8 in. ID Silicone and Pumpsil Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31406.21 | Full specifications | 1 each | Transfer Assembly Pump Tubing, 1/2 in. ID Silicone and Pumpsil Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly Pump Tubing, 1/2 in. ID Silicone and Pumpsil Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31406.31 | Full specifications | 1 each | Transfer Assembly Pump Tubing, 3/4 in. ID Silicone and Pumpsil Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly Pump Tubing, 3/4 in. ID Silicone and Pumpsil Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31407.01 | Full specifications | 1 each | Transfer Assembly Pump Tubing, 3/4 in. ID Silicone and Pumpsil Silicone, AseptiQuik L Connectors | Request A Quote |
Description | Transfer Assembly Pump Tubing, 3/4 in. ID Silicone and Pumpsil Silicone, AseptiQuik L Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31408.01 | Full specifications | 1 each | Transfer Assembly Pump Tubing, 3/8 in. ID Silicone 3/8 in. ID Pumpsil Silicone, MPC Insert and MPC Body | Request A Quote |
Description | Transfer Assembly Pump Tubing, 3/8 in. ID Silicone 3/8 in. ID Pumpsil Silicone, MPC Insert and MPC Body | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|
| SH31409.01 | Full specifications | 1 each | Transfer Assembly Pump Tubing, 3/4 in. ID Silicone and Pumpsil Silicone, MPU Insert and MPU Body | Request A Quote |
Description | Transfer Assembly Pump Tubing, 3/4 in. ID Silicone and Pumpsil Silicone, MPU Insert and MPU Body | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|
| SH31410.01 | Full specifications | 1 each | Transfer Assembly Pump Tubing, 1 in. ID Silicone and Pumpsil Silicone, MPU Insert and MPU Body | Request A Quote |
Description | Transfer Assembly Pump Tubing, 1 in. ID Silicone and Pumpsil Silicone, MPU Insert and MPU Body | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|