Transfer Assembly Pump Tubing
Thermo Scientific™
Transfer Assembly Pump Tubing
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. Simplified connectivity and greater process efficiency for most upstream and downstream process operations.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipos de conectorEstilo de los tubos
SH31414.01Inserto MPC y cuerpo MPC3/8 in DI de silicona trenzada, Pumpsil
SH31314.01Inserto MPU y cuerpo MPU3/4 in TPE y silicona Pumpsil
SH31406.01Conectores AseptiQuik G1/4 in DI de silicona y Pumpsil
SH31406.11Conectores AseptiQuik G3/8 in DI de silicona y Pumpsil
SH31406.21Conectores AseptiQuik G1/2 in DI de silicona y Pumpsil
SH31406.31Conectores AseptiQuik G3/4 in DI de silicona y Pumpsil
SH31407.01Conectores AseptiQuik L3/4 in DI de silicona/Silicona Pumpsil
SH31407.11Conectores AseptiQuik L1 in DI de silicona y Pumpsil
SH31408.01Inserto MPC y cuerpo MPC3/8 in DI de silicona y Pumpsil
SH31409.01Inserto MPU y cuerpo MPU3/4 in DI de silicona y Pumpsil
SH31410.01Inserto MPU y cuerpo MPU1 in DI de silicona y Pumpsil
SH31318.01Inserto MPX y cuerpo MPX1/2 in TPE y silicona Pumpsil
SH31411.01Inserto MPX y cuerpo MPX1/2 in DI de silicona y Pumpsil
SH31320.01Inserto MPC y cuerpo MPC3/8 in TPE y silicona Pumpsil
SH31412.01Conectores AseptiQuik G1/4 in DI de silicona trenzada, Pumpsil
SH31333.01Inserto MPU y cuerpo MPU1 in TPE y silicona Pumpsil
SH31412.11Conectores AseptiQuik G3/8 in DI de silicona trenzada, Pumpsil
SH31341.01Conectores AseptiQuik L3/4 in TPE y silicona Pumpsil
SH31412.21Conectores AseptiQuik G1/2 in DI de silicona trenzada, Pumpsil
SH31341.11Conectores AseptiQuik L1 in TPE y silicona Pumpsil
SH31412.31Conectores AseptiQuik G3/4 in DI de silicona trenzada, Pumpsil
SH31342.01Conectores AseptiQuik G1/4 in TPE y silicona Pumpsil
SH31413.01Conectores AseptiQuik L3/4 in DI de silicona trenzada, Pumpsil
SH31342.11Conectores AseptiQuik G3/8 in TPE y silicona Pumpsil
SH31413.11Conectores AseptiQuik L1 in DI de silicona trenzada, Pumpsil
SH31342.21Conectores AseptiQuik GTPE de 1/2 in y silicona Pumpsil
SH31342.31Conectores AseptiQuik GTPE de 3/4 in y silicona Pumpsil
SH31415.01Inserto MPU y cuerpo MPU3/4 in DI de silicona trenzada, Pumpsil
SH31416.01Inserto MPU y cuerpo MPU1 in DI de silicona trenzada, Pumpsil
SH31417.01Inserto MPX y cuerpo MPX1/2 in DI de silicona trenzada, Pumpsil
Número de catálogo SH31414.01
Precio (CLP)
-
Tipos de conector:
Inserto MPC y cuerpo MPC
Estilo de los tubos:
3/8 in DI de silicona trenzada, Pumpsil
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.
These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.
Especificaciones
Tipos de conectorInserto MPC y cuerpo MPC
DescripciónTransfer Assembly Pump Tubing, 3/8 in. ID Braided Silicone 3/8 in. ID Pumpsil Silicone, MPC Insert and MPC Body
Para utilizar con (aplicación)Monoclonal antibody production, upstream and downstream applications
CantidadEach
Estilo de los tubos3/8 in DI de silicona trenzada, Pumpsil
Unit Size1 each
Contenido y almacenamiento
Room Temperature