Transfer Assembly Jumper Tubing, Braided Silicone, Non-aseptic
Transfer Assembly Jumper Tubing, Braided Silicone, Non-aseptic
Transfer Assembly Jumper Tubing, Braided Silicone, Non-aseptic
Transfer Assembly Jumper Tubing, Braided Silicone, Non-aseptic
Thermo Scientific™

Transfer Assembly Jumper Tubing, Braided Silicone, Non-aseptic

Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. Simplified connectivity and greater process efficiency for most upstream and downstream process operations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesTubing StyleTubing Size
SH31489.22AseptiQuik G Connect, 1 1/2 TriClamp1/2 in. ID Braided Silicone10 ft
SH31223.01Tri-Clamp and MPX1/2 in. ID Braided Silicone5 ft.
SH31396.01MPC Insert and MPC Body3/8 in. ID Braided Silicone5 ft.
SH31403.011 1/2 Tri-Clamp and 3/4 Tri-Clamp1/4 in. ID Braided Silicone5 ft.
SH31223.02Tri-Clamp and MPX1/2 in. ID Braided Silicone10 ft
SH31396.02MPC Insert and MPC Body3/8 in. ID Braided Silicone10 ft
SH31403.021 1/2 Tri-Clamp and 3/4 Tri-Clamp1/4 in. ID Braided Silicone10 ft
SH31223.03Tri-Clamp and MPX1/2 in. ID Braided Silicone15 ft.
SH31396.03MPC Insert and MPC Body3/8 in. ID Braided Silicone15 ft.
SH31403.031 1/2 Tri-Clamp and 3/4 Tri-Clamp1/4 in. ID Braided Silicone15 ft.
SH31225.01Tri-Clamp and MPU Insert1 in. ID Braided Silicone5 ft.
SH31401.01MPX Insert and MPX Body1/2 in. ID Braided Silicone5 ft.
SH31403.111 1/2 Tri-Clamp and 3/4 Tri-Clamp3/8 in. ID Braided Silicone5 ft.
SH31225.02Tri-Clamp and MPU Insert1 in. ID Braided Silicone10 ft
SH31401.02MPX Insert and MPX Body1/2 in. ID Braided Silicone10 ft
SH31403.121 1/2 Tri-Clamp and 3/4 Tri-Clamp3/8 in. ID Braided Silicone10 ft
SH31225.03Tri-Clamp and MPU Insert1 in. ID Braided Silicone15 ft.
SH31401.03MPX Insert and MPX Body1/2 in. ID Braided Silicone15 ft.
SH31403.131 1/2 Tri-Clamp and 3/4 Tri-Clamp3/8 in. ID Braided Silicone15 ft.
SH31402.01MPX Body and 1 1/2 Tri-Clamp1/2 in. ID Braided Silicone5 ft.
SH31403.211 1/2 Tri-Clamp and 3/4 Tri-Clamp1/2 in. ID Braided Silicone5 ft.
SH31402.02MPX Body and 1 1/2 Tri-Clamp1/2 in. ID Braided Silicone10 ft
SH31403.221 1/2 Tri-Clamp and 3/4 Tri-Clamp1/2 in. ID Braided Silicone10 ft
SH31402.03MPX Body and 1 1/2 Tri-Clamp1/2 in. ID Braided Silicone15 ft.
SH31403.231 1/2 Tri-Clamp and 3/4 Tri-Clamp1/2 in. ID Braided Silicone15 ft.
SH31398.01MPU Body and MPU Insert3/4 in. ID Braided Silicone5 ft.
SH31403.311 1/2 Tri-Clamp and 3/4 Tri-Clamp3/4 in. ID Braided Silicone5 ft.
SH31398.02MPU Body and MPU Insert3/4 in. ID Braided Silicone10 ft
SH31403.321 1/2 Tri-Clamp and 3/4 Tri-Clamp3/4 in. ID Braided Silicone10 ft
SH31398.03MPU Body and MPU Insert3/4 in. ID Braided Silicone15 ft.
SH31403.331 1/2 Tri-Clamp and 3/4 Tri-Clamp3/4 in. ID Braided Silicone15 ft.
SH31399.01MPU Insert and 1 1/2 Tri-Clamp3/4 in. ID Braided Silicone5 ft.
SH31404.011 1/2 Tri-Clamps1/4 in. ID Braided Silicone5 ft.
SH31399.02MPU Insert and 1 1/2 Tri-Clamp3/4 in. ID Braided Silicone10 ft
SH31404.021 1/2 Tri-Clamps1/4 in. ID Braided Silicone10 ft
SH31399.03MPU Insert and 1 1/2 Tri-Clamp3/4 in. ID Braided Silicone15 ft.
SH31404.031 1/2 Tri-Clamps1/4 in. ID Braided Silicone15 ft.
SH31398.11MPU Body and MPU Insert1 in. ID Braided Silicone5 ft.
SH31404.111 1/2 Tri-Clamps3/8 in. ID Braided Silicone5 ft.
SH31398.12MPU Body and MPU Insert1 in. ID Braided Silicone10 ft
SH31404.121 1/2 Tri-Clamps3/8 in. ID Braided Silicone10 ft
SH31398.13MPU Body and MPU Insert1 in. ID Braided Silicone15 ft.
SH31404.131 1/2 Tri-Clamps3/8 in. ID Braided Silicone15 ft.
SH31400.01MPU Body and 1 1/2 Tri-Clamp1 in. ID Braided Silicone5 ft.
SH31404.211 1/2 Tri-Clamps1/2 in. ID Braided Silicone5 ft.
SH31400.02MPU Body and 1 1/2 Tri-Clamp1 in. ID Braided Silicone10 ft
SH31404.221 1/2 Tri-Clamps1/2 in. ID Braided Silicone10 ft
SH31400.03MPU Body and 1 1/2 Tri-Clamp1 in. ID Braided Silicone15 ft.
SH31404.231 1/2 Tri-Clamps1/2 in. ID Braided Silicone15 ft.
SH31404.311 1/2 Tri-Clamps3/4 in. ID Braided Silicone5 ft.
SH31404.321 1/2 Tri-Clamps3/4 in. ID Braided Silicone10 ft
SH31404.331 1/2 Tri-Clamps3/4 in. ID Braided Silicone15 ft.
SH31405.011 1/2 Tri-Clamps1 in. ID Braided Silicone5 ft.
SH31405.021 1/2 Tri-Clamps1 in. ID Braided Silicone10 ft
SH31405.031 1/2 Tri-Clamps1 in. ID Braided Silicone15 ft.
SH31489.11AseptiQuik G Connect, 1 1/2 TriClamp3/8 in. ID Braided Silicone5 ft.
SH31489.12AseptiQuik G Connect, 1 1/2 TriClamp3/8 in. ID Braided Silicone10 ft
SH31489.13AseptiQuik G Connect, 1 1/2 TriClamp3/8 in. ID Braided Silicone15 ft.
SH31489.21AseptiQuik G Connect, 1 1/2 TriClamp1/2 in. ID Braided Silicone5 ft.
SH31489.23AseptiQuik G Connect, 1 1/2 TriClamp1/2 in. ID Braided Silicone15 ft.
SH31489.31AseptiQuik G Connect, 1 1/2 TriClamp3/4 in. ID Braided Silicone5 ft.
SH31489.32AseptiQuik G Connect, 1 1/2 TriClamp3/4 in. ID Braided Silicone10 ft
SH31489.33AseptiQuik G Connect, 1 1/2 TriClamp3/4 in. ID Braided Silicone15 ft.
SH31392.01AseptiQuik G Connect, 3/4 Tri-Clamp1/4 in. ID Braided Silicone5 ft.
SH31392.02AseptiQuik G Connect, 3/4 Tri-Clamp1/4 in. ID Braided Silicone10 ft
SH31392.03AseptiQuik G Connect, 3/4 Tri-Clamp1/4 in. ID Braided Silicone15 ft.
SH31392.11AseptiQuik G Connect, 3/4 Tri-Clamp3/8 in. ID Braided Silicone5 ft.
SH31392.12AseptiQuik G Connect, 3/4 Tri-Clamp3/8 in. ID Braided Silicone10 ft
SH31392.13AseptiQuik G Connect, 3/4 Tri-Clamp3/8 in. ID Braided Silicone15 ft.
SH31392.21AseptiQuik G Connect, 3/4 Tri-Clamp1/2 in. ID Braided Silicone5 ft.
SH31392.22AseptiQuik G Connect, 3/4 Tri-Clamp1/2 in. ID Braided Silicone10 ft
SH31392.23AseptiQuik G Connect, 3/4 Tri-Clamp1/2 in. ID Braided Silicone15 ft.
SH31392.31AseptiQuik G Connect, 3/4 Tri-Clamp3/4 in. Braided Silicone5 ft.
SH31392.32AseptiQuik G Connect, 3/4 Tri-Clamp3/4 in. ID Braided Silicone10 ft
SH31394.01AseptiQuik G and MPX Body1/2 in. ID Braided Silicone5 ft.
SH31394.02AseptiQuik G and MPX Body1/2 in. ID Braided Silicone10 ft
SH31394.03AseptiQuik G and MPX Body1/2 in. ID Braided Silicone15 ft.
SH31395.01AseptiQuik L Connect, 1 1/2 TriClamp3/4 in. ID Braided Silicone5 ft.
SH31395.02AseptiQuik L Connect, 1 1/2 TriClamp3/4 in. ID Braided Silicone10 ft
SH31395.03AseptiQuik L Connect, 1 1/2 TriClamp3/4 in. ID Braided Silicone15 ft.
SH31395.11AseptiQuik L Connect, 1 1/2 TriClamp1 in. ID Braided Silicone5 ft.
SH31395.12AseptiQuik L Connect, 1 1/2 TriClamp1 in. ID Braided Silicone10 ft
SH31395.13AseptiQuik L Connect, 1 1/2 TriClamp1 in. ID Braided Silicone15 ft.
SH31392.33AseptiQuik G Connect, 3/4 Tri-Clamp3/4 in. ID Braided Silicone15 ft.
Catalog number SH31489.22
Price (MXN)
-
Connector Types:
AseptiQuik G Connect, 1 1/2 TriClamp
Tubing Style:
1/2 in. ID Braided Silicone
Tubing Size:
10 ft
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.

• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance

Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.

All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.

These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.

Specifications
Connector TypesAseptiQuik G Connect, 1 1/2 TriClamp
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 1/2 in. ID Braided Silicone, AseptiQuik G Connector and 1 1/2 Tri-Clamp
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Tubing Size10 ft
Tubing Style1/2 in. ID Braided Silicone
Unit Size1 each
Contents & Storage
Room Temperature