Transfer Assembly Jumper Tubing, Braided Silicone, Non-aseptic
Transfer Assembly Jumper Tubing, Braided Silicone, Non-aseptic
Transfer Assembly Jumper Tubing, Braided Silicone, Non-aseptic
Transfer Assembly Jumper Tubing, Braided Silicone, Non-aseptic
Thermo Scientific™

Transfer Assembly Jumper Tubing, Braided Silicone, Non-aseptic

Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. Simplified connectivity and greater process efficiency for most upstream and downstream process operations.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipos de conectorEstilo de los tubosTamaño de los tubos
SH31489.32AseptiQuik G Connect, abrazadera triple de 1 1/23/4 in DI de silicona trenzada10 ft
SH31223.01Abrazadera triple y MPX1/2 in DI de silicona trenzada5 ft
SH31396.01Inserto MPC y cuerpo MPC3/8 in DI de silicona trenzada5 ft
SH31403.01Abrazadera triple de 1 1/2 y abrazadera triple de 3/41/4 in DI de silicona trenzada5 ft
SH31223.02Abrazadera triple y MPX1/2 in DI de silicona trenzada10 ft
SH31396.02Inserto MPC y cuerpo MPC3/8 in DI de silicona trenzada10 ft
SH31403.02Abrazadera triple de 1 1/2 y abrazadera triple de 3/41/4 in DI de silicona trenzada10 ft
SH31223.03Abrazadera triple y MPX1/2 in DI de silicona trenzada15 ft
SH31396.03Inserto MPC y cuerpo MPC3/8 in DI de silicona trenzada15 ft
SH31403.03Abrazadera triple de 1 1/2 y abrazadera triple de 3/41/4 in DI de silicona trenzada15 ft
SH31225.01Abrazadera triple e inserto MPU1 in DI de silicona trenzada5 ft
SH31401.01Inserto MPX y cuerpo MPX1/2 in DI de silicona trenzada5 ft
SH31403.11Abrazadera triple de 1 1/2 y abrazadera triple de 3/43/8 in DI de silicona trenzada5 ft
SH31225.02Abrazadera triple e inserto MPU1 in DI de silicona trenzada10 ft
SH31401.02Inserto MPX y cuerpo MPX1/2 in DI de silicona trenzada10 ft
SH31403.12Abrazadera triple de 1 1/2 y abrazadera triple de 3/43/8 in DI de silicona trenzada10 ft
SH31225.03Abrazadera triple e inserto MPU1 in DI de silicona trenzada15 ft
SH31401.03Inserto MPX y cuerpo MPX1/2 in DI de silicona trenzada15 ft
SH31403.13Abrazadera triple de 1 1/2 y abrazadera triple de 3/43/8 in DI de silicona trenzada15 ft
SH31402.01Cuerpo MPX y abrazadera triple 1 1/21/2 in DI de silicona trenzada5 ft
SH31403.21Abrazadera triple de 1 1/2 y abrazadera triple de 3/41/2 in DI de silicona trenzada5 ft
SH31402.02Cuerpo MPX y abrazadera triple 1 1/21/2 in DI de silicona trenzada10 ft
SH31403.22Abrazadera triple de 1 1/2 y abrazadera triple de 3/41/2 in DI de silicona trenzada10 ft
SH31402.03Cuerpo MPX y abrazadera triple 1 1/21/2 in DI de silicona trenzada15 ft
SH31403.23Abrazadera triple de 1 1/2 y abrazadera triple de 3/41/2 in DI de silicona trenzada15 ft
SH31398.01Cuerpo MPU e inserto MPU3/4 in DI de silicona trenzada5 ft
SH31403.31Abrazadera triple de 1 1/2 y abrazadera triple de 3/43/4 in DI de silicona trenzada5 ft
SH31398.02Cuerpo MPU e inserto MPU3/4 in DI de silicona trenzada10 ft
SH31403.32Abrazadera triple de 1 1/2 y abrazadera triple de 3/43/4 in DI de silicona trenzada10 ft
SH31398.03Cuerpo MPU e inserto MPU3/4 in DI de silicona trenzada15 ft
SH31403.33Abrazadera triple de 1 1/2 y abrazadera triple de 3/43/4 in DI de silicona trenzada15 ft
SH31399.01Inserto MPU y abrazadera triple 1 1/23/4 in DI de silicona trenzada5 ft
SH31404.01Abrazaderas triples 1 1/21/4 in DI de silicona trenzada5 ft
SH31399.02Inserto MPU y abrazadera triple 1 1/23/4 in DI de silicona trenzada10 ft
SH31404.02Abrazaderas triples 1 1/21/4 in DI de silicona trenzada10 ft
SH31399.03Inserto MPU y abrazadera triple 1 1/23/4 in DI de silicona trenzada15 ft
SH31404.03Abrazaderas triples 1 1/21/4 in DI de silicona trenzada15 ft
SH31398.11Cuerpo MPU e inserto MPU1 in DI de silicona trenzada5 ft
SH31404.11Abrazaderas triples 1 1/23/8 in DI de silicona trenzada5 ft
SH31398.12Cuerpo MPU e inserto MPU1 in DI de silicona trenzada10 ft
SH31404.12Abrazaderas triples 1 1/23/8 in DI de silicona trenzada10 ft
SH31398.13Cuerpo MPU e inserto MPU1 in DI de silicona trenzada15 ft
SH31404.13Abrazaderas triples 1 1/23/8 in DI de silicona trenzada15 ft
SH31400.01Cuerpo MPU y abrazadera triple 1 1/21 in DI de silicona trenzada5 ft
SH31404.21Abrazaderas triples 1 1/21/2 in DI de silicona trenzada5 ft
SH31400.02Cuerpo MPU y abrazadera triple 1 1/21 in DI de silicona trenzada10 ft
SH31404.22Abrazaderas triples 1 1/21/2 in DI de silicona trenzada10 ft
SH31400.03Cuerpo MPU y abrazadera triple 1 1/21 in DI de silicona trenzada15 ft
SH31404.23Abrazaderas triples 1 1/21/2 in DI de silicona trenzada15 ft
SH31404.31Abrazaderas triples 1 1/23/4 in DI de silicona trenzada5 ft
SH31404.32Abrazaderas triples 1 1/23/4 in DI de silicona trenzada10 ft
SH31404.33Abrazaderas triples 1 1/23/4 in DI de silicona trenzada15 ft
SH31405.01Abrazaderas triples 1 1/21 in DI de silicona trenzada5 ft
SH31405.02Abrazaderas triples 1 1/21 in DI de silicona trenzada10 ft
SH31405.03Abrazaderas triples 1 1/21 in DI de silicona trenzada15 ft
SH31489.11AseptiQuik G Connect, abrazadera triple de 1 1/23/8 in DI de silicona trenzada5 ft
SH31489.12AseptiQuik G Connect, abrazadera triple de 1 1/23/8 in DI de silicona trenzada10 ft
SH31489.13AseptiQuik G Connect, abrazadera triple de 1 1/23/8 in DI de silicona trenzada15 ft
SH31489.21AseptiQuik G Connect, abrazadera triple de 1 1/21/2 in DI de silicona trenzada5 ft
SH31489.22AseptiQuik G Connect, abrazadera triple de 1 1/21/2 in DI de silicona trenzada10 ft
SH31489.23AseptiQuik G Connect, abrazadera triple de 1 1/21/2 in DI de silicona trenzada15 ft
SH31489.31AseptiQuik G Connect, abrazadera triple de 1 1/23/4 in DI de silicona trenzada5 ft
SH31489.33AseptiQuik G Connect, abrazadera triple de 1 1/23/4 in DI de silicona trenzada15 ft
SH31392.01AseptiQuik G Connect, abrazadera triple de 3/41/4 in DI de silicona trenzada5 ft
SH31392.02AseptiQuik G Connect, abrazadera triple de 3/41/4 in DI de silicona trenzada10 ft
SH31392.03AseptiQuik G Connect, abrazadera triple de 3/41/4 in DI de silicona trenzada15 ft
SH31392.11AseptiQuik G Connect, abrazadera triple de 3/43/8 in DI de silicona trenzada5 ft
SH31392.12AseptiQuik G Connect, abrazadera triple de 3/43/8 in DI de silicona trenzada10 ft
SH31392.13AseptiQuik G Connect, abrazadera triple de 3/43/8 in DI de silicona trenzada15 ft
SH31392.21AseptiQuik G Connect, abrazadera triple de 3/41/2 in DI de silicona trenzada5 ft
SH31392.22AseptiQuik G Connect, abrazadera triple de 3/41/2 in DI de silicona trenzada10 ft
SH31392.23AseptiQuik G Connect, abrazadera triple de 3/41/2 in DI de silicona trenzada15 ft
SH31392.31AseptiQuik G Connect, abrazadera triple de 3/43/4 in Silicona trenzada5 ft
SH31392.32AseptiQuik G Connect, abrazadera triple de 3/43/4 in DI de silicona trenzada10 ft
SH31394.01AseptiQuik G y cuerpo MPX1/2 in DI de silicona trenzada5 ft
SH31394.02AseptiQuik G y cuerpo MPX1/2 in DI de silicona trenzada10 ft
SH31394.03AseptiQuik G y cuerpo MPX1/2 in DI de silicona trenzada15 ft
SH31395.01Conector AseptiQuik L, abrazadera triple de 1 1/23/4 in DI de silicona trenzada5 ft
SH31395.02Conector AseptiQuik L, abrazadera triple de 1 1/23/4 in DI de silicona trenzada10 ft
SH31395.03Conector AseptiQuik L, abrazadera triple de 1 1/23/4 in DI de silicona trenzada15 ft
SH31395.11Conector AseptiQuik L, abrazadera triple de 1 1/21 in DI de silicona trenzada5 ft
SH31395.12Conector AseptiQuik L, abrazadera triple de 1 1/21 in DI de silicona trenzada10 ft
SH31395.13Conector AseptiQuik L, abrazadera triple de 1 1/21 in DI de silicona trenzada15 ft
SH31392.33AseptiQuik G Connect, abrazadera triple de 3/43/4 in DI de silicona trenzada15 ft
Número de catálogo SH31489.32
Precio (MXN)
-
Tipos de conector:
AseptiQuik G Connect, abrazadera triple de 1 1/2
Estilo de los tubos:
3/4 in DI de silicona trenzada
Tamaño de los tubos:
10 ft
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.

• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance

Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.

All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.

These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.

Especificaciones
Tipos de conectorAseptiQuik G Connect, abrazadera triple de 1 1/2
DescripciónTransfer Assembly Jumper Tubing, 10 ft., 3/4 in. ID Braided Silicone, AseptiQuik G Connector and 1 1/2 Tri-Clamp
Para utilizar con (aplicación)Monoclonal antibody production, upstream and downstream applications
CantidadEach
Tamaño de los tubos10 ft
Estilo de los tubos3/4 in DI de silicona trenzada
Unit Size1 each
Contenido y almacenamiento
Room Temperature