Self Collection Kit for Nasal Samples
Thermo Scientific™
Self Collection Kit for Nasal Samples
Rapidly inactivate and stabilize viral RNA at ambient temperature with a non-hazardous formulation using this Self Collection Kit for Nasal Samples
Have Questions?
Número do catálogoQuantity
SK0137320
Número do catálogo SK01373
Preço (BRL)
-
Quantity:
20
Rapidly inactivate and stabilize viral RNA at ambient temperature with a non-hazardous formulation using Thermo Scientific™ Self Collection Kit for Nasal Samples. This kit is intended for self-collection of nasal samples
Safely collect and inactivate RNA enveloped viruses with Thermo Scientific™ Self Collection Kit for Nasal Samples. The viral inactivation meduim stabilises the viral RNA for transportation and use in subsequent in vitro diagnostic testing procedures.
The Thermo Scientific™ Self Collection Kit for Nasal Samples includes a convenient, leakproof format that can be used with unprocessed nasal swabs, and the non-hazardous formulation enables safe transport of samples and is suitable for self-collection use.
Each Thermo Scientific™ Self Collection Kit for Nasal Samples comes ready to use with individually packaged kits which contain the required materials for self collection: a 10mL tube filled with InhibiSURE Viral Inactivation Media, a single-use only nasal swab, return packaging and associated labels.
To use the kit, the individual follows 5 easy general steps:
  • Check kit components and read the instructions for use prior to sample collection
  • Wash your hands
  • Collect the nasal sample with the swab provided and place sample into the tube of liquid medium
  • Wash your hands
  • Package and prepare the sample for postage
The device is intended for self-collection of nasal samples. For professional, non-laboratory, and self-collection use. This is a single use device. This product is suitable for use by: Adults aged 18 and over. Ages 14 - 17 under supervision by an individual over 18. Samples for children under the age of 13 years should be collected by an individual over 18.
Product is CE marked but not 501(k) cleared and not available for sale in the U.S. Availability of product in each country depends on local regulatory marketing status.
For In Vitro Diagnostic Use.  
Especificações
Closure ColorPurple
CompatibilityInhibiSURE™ Viral Inactivation Medium has been designed to be compatible with nucleic acid purification and amplification workflows. It has been validated with the Thermo Scientific™ MagMax™ MVP II kit (product code A48383) and TaqPath™ COVID 19 CE IVD RT PCR Kit (A48067)
DescriptionThermo Scientific™ Self Collection Kit for Nasal Samples
Fill Volume1.5 mL
For Use With (Application)Medium is a liquid medium intended to be used for the collection and inactivation of RNA enveloped viruses. The product stabilises the viral RNA for transportation and use in subsequent in vitro diagnostic testing procedures. The device is intended for self-collection of nasal samples. For professional, non-laboratory, and self-collection use. This is a single use device. This product is suitable for use by: Adults aged 18 and over. Ages 14 - 17 under supervision by an individual over 18. Samples for children under the age of 13 years should be collected by an individual over 18.
Quantity20
Storage Requirements2 - 30°C
Transport Temperature2 - 30°C
Unit SizeEach